missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BIX NHE1 inhibitor
GREENER_CHOICE

Product Code. p-7110838
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18738229

Brand: Tocris Bioscience™ 5512/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective NHE1 inhibitor

BIX NHE1 inhibitor is a potent and selective NHE1 inhibitor (IC50 = 31 nM). Selective for NHE1 over NHE2 and NHE3. Prevents ischemic damage in an ischemia reperfusion injury isolated rat heart model ex vivo. Prevents phenylephrine -induced cardiomyocyte hypertrophy in vitro, and attenuates cardiac hypertrophy and left ventricular dysfunction postinfarction in rats. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide hydrochloride
CAS 1422252-46-7
Quantity 10 mg
Target Na+/Ca2+ Exchanger Inhibitors
Molecular Formula C16H19F3N4O2.HCl
Formula Weight 392.8
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.