missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ BK Polyomavirus Antigen Microarray
Description
- Microarray Content: Small T antigen, Large T antigen, Major capsid proteins VP1, VP2, and VP3
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 1836 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
Specifications
| Coating | PEGMA/PMMA |
| Description | 2 array copies per microarray |
| Quantity | 1 Slide |
| Method of Attachment | Combinatorial peptide synthesis by amino acid laser printing |
| Thickness (Metric) | 1 mm |
| Certifications/Compliance | ISO 9001:2015 |
| Color | Transparent |
| Regulatory Status | RUO |
| Barcode | Individual barcode for each microarray |
| Material | Microscopic Glass |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?