missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
BIOPHARMA

Product Code. 15541413 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 3840
Pack of 5760
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Unit Size
15541413 300 μL Sterile Filtered Pack of 5760
15551393 10 μL Sterile Non-filtered Pack of 5760
15541393 10 μL Non-sterile Non-filtered Pack of 5760
15501413 10 μL Sterile Filtered Pack of 5760
15571403 50 μL Non-sterile Non-filtered Pack of 5760
15581403 50 μL Sterile Non-filtered Pack of 5760
15561413 50 μL Sterile Filtered Pack of 5760
15531403 300 μL Sterile Non-filtered Pack of 5760
15581393 1000 μL Non-sterile Non-filtered Pack of 3840
15521413 1000 μL Sterile Filtered Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15541413 Supplier Axygen™ Supplier No. HTF300CBKHTRS5
  • ONLINE EXCLUSIVE PRICE
    901.00 €
    745.44 € / Pack of 5760
    Save 155.56 €
    17% Off
Estimated Shipment: 10-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations

Axygen offers an alternative choice for your Hamilton™ STARline system.

These tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation, Round
Packaging 5 Racks/Pk, 12 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.