missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Black Acrylonitrile-Butadiene-Rubber Tubing

Providing oil resistance in the laboratory.

Brand:  Deutsch Neumann™ 3240408

 View more versions of this product

Product Code. 10014980

  • 62.20 € / 25m
Only null left
Estimated Shipment: 06-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Non-reinforced
  • Acrylonitrile-Butadiene-Rubber
  • Minimum quantity of 5m
  • Available in a variety of inner diameters

Specifications

Specifications

ISO 9001:2008
NBR
5 m
Oil resistant
4 mm
8 mm
2 mm
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.