Learn More
Fisherbrand™ Black Phenolic Caps
Product Code. p-7144783 Shop All Fisherbrand Products
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Unit Size:
Pack of 1600
Pack of 3100
Pack of 5500
Pack of 576
Product Code. 16174397
Only null left
Add to basket
Add to basket
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Color Black
Quantity 1600
Closure Color Black
Neck Finish 38-400
Closure Included No
Closure Liner Polyethylene Cone
Closure Material Phenolic