missing translation for 'onlineSavingsMsg'
Learn More

WHEATON™ Black Screw Cap

Product Code. 30286516 Shop All Wheaton Science Products Products
Change view
Click to view available options
Quantity:
140/Pk.
190/Pk.
231/Pk.
264/Pk.
399/Pk.
666/Pk.
882/Pk.
Unit Size:
Pack of 140
Pack of 190
Pack of 231
Pack of 264
Pack of 399
Pack of 666
Pack of 882
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286516 264/Pk. Pack of 264
30286523 666/Pk. Pack of 666
30286506 882/Pk. Pack of 882
30286528 399/Pk. Pack of 399
30286513 231/Pk. Pack of 231
30286511 190/Pk. Pack of 190
30286515 140/Pk. Pack of 140
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30286516 Supplier WHEATON™ Supplier No. 110062264
Add to basket
Edge
Add to basket

Tubular Dram Vial Caps

  • WHEATON™ tubular dram vials are manufactured from clear 5.1 expansion borosilicate glass and use WHEATON™ black polypropylene caps with foil liner
  • Both vials and caps are suitable for storage, display, sample collection and apothecary applications
  • The vials have a buttress thread finish and fit with the buttress thread black polypropylene dram vial caps with foil liner and range from 1.75 mL to 28 mL in volume
  • Dram vial caps are available in black only and are manufactured from polypropylene and have a foil liner, and provide good barrier properties and resistance to hydrocarbons, oils, ketones and alcohol
  • Black polypropylene foil lined dram vial caps are sold separately to match vials sizes and quantity per pack
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Diameter (Metric) Outer 18 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Quantity 264/Pk.
For Use With (Equipment) Vial
Autoclavable Autoclavable
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.