missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Black Urea Screw Caps with Wood Pulp Faced PVDC Liner

Product Code. p-8001884
Click to view available options
Neck Finish:
100/R3
24/R3
28/R3
31/R6
33/R3
38/R3
48/R3
51/R3
58/R3
63/R3
83/R3
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 12397048

Brand: Azpack™ 12397048

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Creates a tight, leakproof seal. Azpack™ Black Urea Screw Caps with Wood Pulp Faced PVDC Liner protect against harsh chemicals.

  • Black urea plastic
  • Includes fitted wood pulp faced PVDC liner
  • Supplied in packs of 50

Specifications

Color Black
Material Urea
Quantity 50/PK
Liner Material Wood pulp faced PVDC
Neck Finish 24/R3
Product Type Screw Cap
For Use With (Equipment) Wide mouth bottles and jars
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.