missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Blind stopper for load carrier

Product Code. 10120102 Shop All Miele Products
Change view
Click to view available options
Product Type:
Blanking Screw
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10120102 Blanking Screw Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10120102 Supplier Miele Professional™ Supplier No. 3809630
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Blind stopper E 362 for the sealing of fittings to load carriers

Thread M 8 x 1
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Blanking Screw
Color Stainless Steel
Model E 362
For Use With (Equipment) Washer-disinfectors, laboratory
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.