missing translation for 'onlineSavingsMsg'
Learn More

Blarcamesine hydrochloride, MedChemExpress

Product Code. 16493618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493618 10 mg 10mg
16474706 100 mg 100mg
16466347 5 mg 5mg
16430017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493618 Supplier Medchem Express Supplier No. HY101864
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.

Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.

Specifications

Chemical Name or Material Blarcamesine hydrochloride
CAS 195615-84-0
Color White
Purity Grade Notes Research
Molecular Formula C19H24ClNO
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (314.61 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (78.65 mM; Need ultrasonic)
SMILES CN(C)CC1CCOC1(C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl
Molecular Weight (g/mol) 317.85
Formula Weight 317.85
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.