missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling

Product Code. 10450751 Shop All Brand Products
Change view
Click to view available options
Includes:
With spring clips
Without spring clips
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10450751 With spring clips Each
10360141 Without spring clips Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10450751 Supplier BRAND™ Supplier No. 718620
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.

  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • The ruling shows 9 large squares of 1 mm2 each.
  • The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.
  • The large square in the center is subdivided into 16 squares with 0.25 mm sides.
  • Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm2.
  • The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.
  • Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Counting Device
Quantity 1Each
Depth (Metric) 0.1mm
Certifications/Compliance DIN EN 285
For Use With (Application) Counting leucocytes, thrombocytes and erythrocytes
Includes With spring clips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.