missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (-)-Blebbistatin

Product Code. 17150062
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17150062 5 mg 5mg
17160062 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150062 Supplier STEMCELL Technologies Supplier No. 72402
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Non-muscle myosin II (NM II) ATPase inhibitor

  • (-)-Blebbistatin is a selective cell-permeable inhibitor of non-muscle myosin II ATPases (Kovács et al.; Straight et al.) It rapidly and reversibly inhibits Mg-ATPase activity and in vitro motility of non-muscle myosin IIA and IIB for several species (IC50 = 0.5-5.0 μM), while poorly inhibiting smooth muscle myosin (IC50 = 80 μM) (Limouze et al.)
  • Increases human pluripotent stem cell (hPSC) survival and cloning efficiency after dissociation to single cells, downstream of ROCK inhibition (Chen et al.; Ohgushi et al.; Walker et al.; Xu et al.)
  • Enables hPSC to be cultured on microcarriers without surface coating (Chen et al.)
  • Inhibits differentiation of human mesenchymal stem cells (McBeath et al.; Engler et al.)

Specifications

CAS 856925-71-8
Quantity 5 mg
Molecular Formula C18H16N2O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.