missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (±)-Blebbistatin

Product Code. 30002125
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002125 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30002125 Supplier R&D Systems™ Supplier No. 1760/10
  • ONLINE EXCLUSIVE PRICE
    477.79 €
    428.40 € / 10mg
    Save 49.39 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of myosin II ATPase activity

(±)-Blebbistatin is a selective inhibitor of myosin II ATPase activity; inhibits contraction of the cleavage furrow without disrupting mitosis or contractile ring assembly. (?)-Blebbistatin rapidly and reversibly blocks cell blebbing, and disrupts directed cell migration and cytokinesis in vertebrate cells. (?)-Blebbistatin inhibits arterial thrombosis development, reduces infiltration of inflammatory cells, and reduces damage to vascular tissue in vivo in a mice model of arterial thrombosis.

Active and Inactive Enantiomer also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 674289-55-5
Quantity 10 mg
Product Line Tocris
Molecular Formula C18H16N2O2
Formula Weight 292.34
Purity >98%
Molecular Weight (g/mol) 292.338
Chemical Name or Material (±)-Blebbistatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.