missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bleomycin sulfate

Product Code. 16457398
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457398 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457398 Supplier MedChemExpress Supplier No. HY17565
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bleomycin sulfate is a DNA synthesis inhibitor. Bleomycin hydrochloride is a DNA damaging agent. Bleomycin sulfate is an antitumor antibiotic.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 1512.62 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 9041-93-4
Product Type Natural Products
Purity 95.0%
Solubility H2O : 255 mg/mL (168.58 mM∣Need ultrasonic and warming) ∣DMSO : 62.5 mg/mL (41.32 mM∣Need ultrasonic)
Target DNA/RNA Synthesis∣Antibiotic
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.