missing translation for 'onlineSavingsMsg'
Learn More

Bleomycin Sulfate, MP Biomedicals™

Product Code. 15779150 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15779150 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779150 Supplier MP Biomedicals Supplier No. 0219030610
  • ONLINE EXCLUSIVE PRICE
    599.00 €
    568.05 € / 10mg
    Save 30.95 €
    5% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bleomycin Sulfate from Streptomyces verticillus, White powder

  • It is an atineoplastic antibiotic.
  • A mixture of cytotonic glycopeptide antibiotics which reacts with DNA and causes strand scission; they have also been shown to have a type of oxygen transferase activity. Bleomycin A2 is the main component of the bleomycin employed clinically.
  • Bleomycins differ from one another in the terminal amine and show varying biological activity.

Specifications

CAS 9041-93-4
Color White
Molecular Formula C55H85N17O25S4
MDL Number MFCD00070310
Quantity 10 mg
Synonym Blenoxane, Blexane, Bleo
Solubility Information Very soluble in water, methanol; slightly soluble in ethanol. Practically insoluble in acetone, ethyl acetate, butyl acetate, ether.
InChI Key WUIABRMSWOKTOF-OCBSMOPSSA-N
Molecular Weight (g/mol) 1512.62
PubChem CID 122172957
Grade >99.0%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.