missing translation for 'onlineSavingsMsg'
Learn More

SCAT Blind Plugs for HPLC

Product Code. 10226504 Shop All SCAT Europe Products
Change view
Click to view available options
For Use With:
Exhaust Filter
Ground Neck Bottles w/ Locknut
Safety Caps, Safety Waste Caps
Color:
Colorless
White
Material:
PFA
PTFE
Thread Size:
GL14
NPT 1/8 in.
NS29/32
UNF 1/4 in. 28G
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Color Material Thread Size unitSize
10226504 Exhaust Filter White PTFE GL14 Each
13233859 Safety Caps, Safety Waste Caps Colorless PFA UNF 1/4 in. 28G Pack of 10
10051453 Safety Caps, Safety Waste Caps Colorless PTFE NPT 1/8 in. Each
10357579 Ground Neck Bottles w/ Locknut White PTFE NS29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10226504 Supplier SCAT Supplier No. 107620
  • ONLINE EXCLUSIVE PRICE
    29.62 €
    25.27 € / Pack of 1
    Save 4.35 €
    14% Off
View all promotions
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Blind plugs for use with HPLC instruments.

Be prepared for HPLC with these SCAT™ blind plugs.

Specifications

Type Blind Plug
Color White
For Use With (Application) HPLC/UHPLC Systems
Thread Size GL14
For Use With Exhaust Filter
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.