missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLOCK-iT™ Fluorescent Oligo, for electroporation
Click to view available options
Quantity:
75 μL
Unit Size:
Each
Description
BLOCK-iT™ Fluorescent Oligo provides:
A strong, easily detectable FITC signal indicating transfection efficiency
Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA
Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
This makes the BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. BLOCK-iT™ Fluorescent Oligo is available in two configurations: one designed for lipid-mediated transfection (2 x 125μL) at 20μM and one designed for electro-poration (75μl) at 1 mM.
This makes the BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. BLOCK-iT™ Fluorescent Oligo is available in two configurations: one designed for lipid-mediated transfection (2 x 125μL) at 20μM and one designed for electro-poration (75μl) at 1 mM.
Specifications
Specifications
| Content And Storage | The BLOCK-iT™ Fluorescent Oligo for electroporation is supplied as 75 μl at 1 mM, and should be stored at -20° C. |
| Control Type | Transfection Control |
| Detection Method | Fluorescence |
| Sample Type | RNA |
| For Use With (Application) | Electroporation |
| Label or Dye | FITC (Fluorescein) |
| Product Line | BLOCK-iT |
| Product Type | Fluorescent Oligo |
| Quantity | 75 μL |
| RNAi Type | siRNA |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction