missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Product Code. 18643873 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Chemical Name or Material:
PBS
TBS
Unit Size:
1 L
100mL
Pack of 3
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material unitSize
18643873 3 x 1 L PBS Pack of 3
13454299 100 mL PBS 100mL
10496134 1 L PBS 1 L
13464299 100 mL TBS 100mL
10443644 1 L TBS 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18643873 Supplier Thermo Scientific™ Supplier No. 37528X3
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:

  • Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
  • Easy to use—1% casein solution is ready to use; can be diluted further as needed
  • Safe—stable, thimerosal-free formulation
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PBS
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Blocker
Quantity 3 x 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.