missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BloodStor™ 100

Product Code. p-7217765
Encompass_Preferred
Click to view available options
Quantity:
50 mL
100 mL
5 x 100 mL
Unit Size:
100mL
50mL
Pack of 5
This item is not returnable. View return policy

Product Code. 17129931

Brand: STEMCELL Technologies 07951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biopreservation reagent for cells and tissues

  • BloodStor™ is a series of generic cGMP freezing media products used to cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics
  • BloodStor™ 100 contains 100% (w/v) DMSO USP

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cryopreservation
Quantity 50 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.