missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 2 grade is ideal for gel wicking and drying, capillary blotting using western, Southern, or semidry techniques.
Specifications
Specifications
| Description | Capillary rise: 70mm/10 min., Weight: 195g/m2, 100/Box |
| Dimensions (L x W) | 130 x 120 mm |
| Film Thickness | 0.35 mm |
| Grade | BF 2 |
| Width (Metric) | 120 mm |
| Length (Metric) | 130 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?