missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Discs

Product Code. 15942960 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Outer:
22 mm
25 mm
150 mm
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
15942960 150 mm Pack of 100
15922960 22 mm Pack of 1000
15932960 25 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15942960 Supplier Sartorius Stedim Biotech™ Supplier No. FT3520150
  • ONLINE EXCLUSIVE PRICE
    1120.00 €
    792.11 € / Pack of 100
    Save 327.89 €
    29% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3 Discs are highly pure and ideal for blotting chromatography.

  • This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Made of high-purity cotton linters for uniform buffer flow and resulting blots.
  • No additives to avoid any interference during transfer.
  • BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
TRUSTED_SUSTAINABILITY

Specifications

Description Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk
Diameter (Metric) Outer 150 mm
Film Thickness 0.76 mm
Format Disc
Grade BF 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.