missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets

Product Code. 15992800 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
135 mm
200 mm
Width (Metric):
155 mm
200 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
15992800 200 mm 200 mm Pack of 100
15982800 135 mm 155 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15992800 Supplier Sartorius Stedim Biotech™ Supplier No. FT2520200200N
  • ONLINE EXCLUSIVE PRICE
    161.00 €
    113.68 € / Pack of 100
    Save 47.32 €
    29% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets are highly pure and ideal for blotting chromatography.

  • This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Made of high-purity cotton linters for uniform buffer flow and resulting blots.
  • No additives to avoid any interference during transfer.
  • BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
TRUSTED_SUSTAINABILITY

Specifications

Description Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk
Dimensions (L x W) 200 x 200 mm
Film Thickness 0.76 mm
Format Sheet
Grade BF 3
Width (Metric) 200 mm
Length (Metric) 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.