missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Blu10 Prestained Protein Ladder

Product Code. 30176230
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30176230 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30176230 Supplier R&D Systems™ Supplier No. NBP333171
  • ONLINE EXCLUSIVE PRICE
    222.01 €
    170.10 € / 500µL
    Save 51.91 €
    23% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Prestained Protein Ladder

The Blu10 Prestained Protein Ladder is a combination of 10 pre-stained proteins with molecular weights from 11 to 180 kDa. The 10 recombinant proteins are covalently coupled with blue chromophore, while 1 green band at 25 kDa and a red band at 75 kDa, serve as reference bands. The Blu10 Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating protein size, and validating Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
TRUSTED_SUSTAINABILITY

Specifications

Formulation 20 mM Trisphosphate(pH 7.5), 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15% (v/v) Glycerol.
For Use With (Application) 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring.
Quantity 500 μL
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.