Learn More
Novus Biologicals™ Blu11 Prestained Protein Ladder
Product Code. 30161595
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Product Code. Quantity unitSize
30161595 500 μL 500µL
1 options
Product Code. 30161595 Supplier Novus Biologicals™ Supplier No. NBP333172
  • ONLINE EXCLUSIVE PRICE
    191.00 €
    180.60 € / 500µL
    Save 10.40 €
    5% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Prestained Protein Ladder
The Blu11 Prestained Protein Ladder is a combination of 11 pre-stained proteins with molecular weights from 10 to 180 kDa. The 11 recombinant proteins are covalently coupled with blue chromophore, and 2 reference bands at 25 kDa and 72 kDa serve as reference bands. The Blu11 Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating protein size and validating Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
TRUSTED_SUSTAINABILITY
Specifications
Formulation 20 mM Trisphosphate (pH 7.5) at 25C, 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15% (v/v) Glycerol.
For Use With (Application) 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring.
Quantity 500 μL