Learn More
Description
Thermo Scientific™ Blue Carrier Protein is a highly-soluble, mollusk-derived hemocyanin formulated in convenient package sizes and a complete kit for carbodiimide (EDC-mediated) hapten-carrier conjugation via amine and carboxyl groups.
Highlights:
Convenient packages – choose lyophilized Blue Carrier Protein or complete EDC conjugation kit
High-yield conjugation – each molecule of CCH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers
Validated quality – the immunogenic carrier protein is specially purified to ensure uniform chemical structure and solubility for consistent performance
Superior solubility – increased solubility compared to KLH allows Blue Carrier Protein to be used in a broader range of conjugation protocols to accommodate unusual or poorly soluble haptens
Highly immunogenic – provides essentially the same level of immunogenicity and phylogenetic distance as keyhole limpet hemocyanin (KLH)
Properties of Blue Carrier Protein:
- Alternative names: Concholepas concholepas hemocyanin, CCH
- Molecular weight: Whole molecule: ∽8,000,000; Subunits: ∽400,000 and ∽350,000
- Absorption: maxima 280nm, 350nm and 558nm
- Extinction coefficient (Abs. 1mg/mL solution): 1.4 at 280nm; 0.35 at 346nm
- A280/A350 Ratio: ∽4.0
- Copper/protein ratio: ∽16 Cu2+/protein molecule
- Isoelectric point: 6.0
- Solubility: Soluble in aqueous buffers pH 7.0 to 8.0
- Stability: Stable in aqueous buffers (∽pH 7.2 without Ca2+ or Mg2+)
Includes:
Kit contains lyophilized Blue Carrier Protein (in MES buffer), EDC crosslinker, buffers and desalting columns to easily prepare ready-to-use immunogens for injection
Specifications
Specifications
| Content And Storage | 4°C. Shipped on ice. |
| Quantity | 500 mg |
| Product Line | Imject |
| Product Type | Imject Blue Carrier Protein |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.