Learn More
SHOWA™ Blue Chemical Protection Gloves, Model CS700
Product Code. 17817351
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
Product Code. 17817351
Brand: SHOWA™ CS70008M
Only null left
Add to basket
Edge
Add to basket
With anti-slip technology, the CS700 food-safe gloves offer ice-grip and increased tactility.
  • A double-dipped nitrile coating provides chemical protection and ensures the gloves are highly durable to keep the wearer safe from harmful substances.
  • The combination of a seamless liner and natural latex-free design is kind to the skin.
Specifications
Chlorinated No
Color Blue
Finger Thickness (English) 45.6 mil
Finger Thickness (Metric) 1.16 mm
Latex Free Yes
Material Nitrile, Polyester
Palm Thickness (English) 45.6 mil
Palm Thickness (Metric) 1.16 mm
Size 8
Thickness (English) 45.6 mil
Thickness (Metric) 1.16 mm
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMNOPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Lining Lined