missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Blue Clickmaster Ratchet Nut for FastPrep-24™ 5G and Classic

Product Code. 17936854 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-245G and Classic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17936854 FastPrep-245G and Classic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17936854 Supplier MP Biomedicals™ Supplier No. 116004525
Add to basket
Add to basket
This item is not returnable. View return policy

Supports up to 11 different adapters. From light weighted ones to heavy and sturdy adapters, the Blue Clickmaster provides the easiest and most secure locking mechanism.

  • The FastPrep™ is the only available homogenizer with multiple interchangeable adapters which allows a unique versatility in sample number, size, and temperature conditions. All optional adapters stand stable on the benchtop and are commonly used as tube racks for sample storage at -20°C or -80°C.
  • The securing knob is important for maintaining the tube holder adapters on the central axis or during the 3-step assembly of the FastPrep-24™.
  • One Blue Clickmaster Ratchet Nut supports up to 11 different adapters. From lightweight ones to heavy and sturdy adapters, the Blue Clickmaster provides the easiest and most secure locking mechanism. Thanks to its high torque, it is particularly recommended for the heaviest adapters and especially for metal tube holders.
  • This product can be used on FastPrep-24™Classic or FastPrep-24™ 5G
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ratchet Nut
Blade Material Stainless Steel
Closure Color Blue
For Use With (Equipment) FastPrep-245G and Classic

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.