missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Blue Dextran 2000 Protein Standard

Product Code. 11574895 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
11574895 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 11574895

Brand: Cytiva 17036001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Determine the void fraction in high-resolution gel filtration columns

  • For use in protein isolation and purification

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Form Solid
For Use With (Application) Determine the void volume in Gel filtration columns
Quantity 10 g
Product Type Protein Molecular Weight Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.