Learn More
3M™ Protective Coverall 4532+, Blue
Product Code. 15720048
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15720048 Large Pack of 20
15780038 Small Pack of 20
15710048 Medium Pack of 20
15750038 X-Large Pack of 20
16314658 2X-Large Pack of 20
15730048 3X-Large Pack of 20
15750048 4X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15720048 Supplier 3M™ Supplier No. 7100015994
Only null left
Add to basket
Add to basket
Coverall has a unique specialist material treatment which offers enhanced liquid hold out with added oil and alcohol repellency. Certified to Type 5 for hazardous particles and Type 6 for limited chemical splash, 4532+ coverall offers the soft breathable comfort of an SSMMS with the improved liquid performance normally associated with a laminate.
  • Unique liquid repellency with oil and alcohol resistance material treatment
  • Soft and strong multi-layered 52 gsm SSMMS for strong performance
  • Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Large
Gender Unisex
Material SSMMS
Color Blue
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Closure Type Front Zipper
Sterility Non-sterile
Certifications/Compliance CE Category III , EN 1073-2 radioactive particles , EN 1149-5 - electrostatic properties , EN 13034: 2005 + A1: 2009 Type 6 liquid splash , EN ISO 13982-1: 2004 + A1: 2010 Type 5 harmful particles
Certifications/Compliance (Europe) CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5