missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-coded Micro PTFE Stir Bar

Product Code. 12696316 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
2 mm
5 mm
7 mm
8 mm
10 mm
15 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Color:
Blue
Red
Yellow
Unit Size:
Pack of 10
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Color unitSize
12696316 5 mm 2 mm Red Pack of 10
11517193 15 mm 1.5 mm Blue Pack of 10
11597183 8 mm 1.5 mm Blue Pack of 10
11567183 2 mm 2 mm Blue Pack of 10
11577183 5 mm 2 mm Blue Pack of 10
11587183 7 mm 2 mm Blue Pack of 10
12686316 10 mm 3 mm Blue Pack of 10
11567193 15 mm 1.5 mm Yellow Pack of 10
11557193 10 mm 3 mm Yellow Pack of 10
11547193 7 mm 2 mm Yellow Pack of 10
11537193 5 mm 2 mm Yellow Pack of 10
11527193 2 mm 2 mm Yellow Pack of 10
11537183 8 mm 1.5 mm Red Pack of 10
11557183 15 mm 1.5 mm Red Pack of 10
11547183 10 mm 3 mm Red Pack of 10
11527183 7 mm 2 mm Red Pack of 10
11507183 2 mm 2 mm Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 12696316
Only null left
Add to basket
Add to basket

Use when quick visual identification is highly important. Fisherbrand™ Color-coded Micro PTFE Stir Bar is available in three colors: blue, red, and yellow.

  • Color-coded, micro stirrer bars
  • For use where identification is of prime importance
  • Note: Colored PTFE coatings are not as inert as pure PTFE

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Red
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 2 mm
Length (Metric) 5 mm
Quantity 10/Pk.
Shape Micro
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.