missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Blue Sepharose™ 6 Fast Flow Chromatography Media

Product Code. 10271350 Shop All Cytiva Products
Change view
Click to view available options
Particle Size:
45 to 165 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
10271350 45 to 165 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10271350 Supplier Cytiva Supplier No. 17094801
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Blue dye binds to proteins such as albumin and lipoproteins as well as enzymes like kinases and dehydrogenases

This medium is well established as adsorbent for albumin and interferon at laboratory and process scale.

  • Cibacron Blue 3G is covalently coupled to the base matrix allowing for low ligand leakage and high capacity
  • The blue dye binds proteins, such as albumin, interferon, lipoproteins and blood coagulation factors
  • The dye also binds enzymes including kinases, dehydrogenases, and most enzymes requiring adenyl-containing cofactors e.g. NAD+
  • Avg. particle size: 90μm
  • Volume: 50mL

Developed and tested in co-operation with large-scale manufacturers and is used in routine commercial production.

  • For large-scale purification of interferon and albumin
  • For isolating groups of enzymes and removing of albumin
  • For industrial applications and other approved applications

Certifications

Certificate of Analysis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Stability Stable in commonly used aqueous Buffers: 70% Ethanol, 6 M Guanidine Hydrochloric, 8 M Urea
For Use With (Application) For Albumin and Interferon at Both Laboratory and Process Scale
Storage Requirements 2°C to 8°C
Product Type Blue Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Includes Certificate of Analysis
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 4 to 12
Storage Buffer 20% Ethanol, 0.1 M Potassium Phosphate Buffer
Ligand Type Cibacron Blue
Show More Show Less
compliance-icons
Product Identifier
  • Blue Sepharose 6 Fast Flow, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-1.2 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 16.5% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.