Learn More
Beschreibung
- Our range of single use, disposable liquid funnels are ideal for use in laboratories and manufacturing areas.
- These funnels are made from food grade polymers and are designed to be used straight from the bag.
- The bright blue color means they can be easily seen if they drop in or left in the material by mistake.
- SteriWare™ Funnels are available sterile and non-sterile.
- SteriWare™ is the largest range of single-use equipment for sampling, laboratory and production work on the market.
- The whole range is made in a cleanroom and comes with class-leading pharmaceutical documentation.
- Simply unwrap the funnel and use.
- The funnels come with a side tab so they are easy to hold.
- The individually bagged funnels are made from PP that is FDA/EC compliant.
- The material used is also certified BSE/TSE free. The sterile funnels conform to SAL 10-6
- The funnels come with a batch-specific certificate that includes the date of manufacture, expiry date and irradiation certificate (for sterile funnels only).
Spezifikation
Spezifikation
| Diameter (Metric)Top | 100 mm |
| Material | Polypropylene |
| Color | Blue |
| Sterility | Non-sterile |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?