Learn More
Sampling Systems™ Blue SteriWare™ Liquid Funnels
BIOPHARMA
Product Code. 17584656 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
17584656 Sterile Pack of 50
17574656 Non-sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17584656 Supplier Sampling Systems™ Supplier No. 8370B100S
Only null left
Add to basket
Add to basket
Remove the risk of cross contamination and improve your quality control practices by dispensing liquids with blue SteriWare™ Liquid Funnels. Manufactured in our cleanroom and available sterile, these funnels are ideal for use in a wide range of applications in the food industry.
  • Our range of single use, disposable liquid funnels are ideal for use in laboratories and manufacturing areas.
  • These funnels are made from food grade polymers and are designed to be used straight from the bag.
  • The bright blue color means they can be easily seen if they drop in or left in the material by mistake.
  • SteriWare™ Funnels are available sterile and non-sterile.
  • SteriWare™ is the largest range of single-use equipment for sampling, laboratory and production work on the market.
  • The whole range is made in a cleanroom and comes with class-leading pharmaceutical documentation.
  • Simply unwrap the funnel and use.
  • The funnels come with a side tab so they are easy to hold.
  • The individually bagged funnels are made from PP that is FDA/EC compliant.
  • The material used is also certified BSE/TSE free. The sterile funnels conform to SAL 10-6
  • The funnels come with a batch-specific certificate that includes the date of manufacture, expiry date and irradiation certificate (for sterile funnels only).
Specifications
Diameter (Metric)Top 100 mm
Material Polypropylene
Color Blue
Sterility Sterile