Learn More
Sampling Systems™ Blue SteriWare™ Scrapers
BIOPHARMA
Product Code. 17594686 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
17594686 Sterile Pack of 100
17584686 Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17594686 Supplier Sampling Systems™ Supplier No. 8785B75S
  • ONLINE EXCLUSIVE PRICE
    205.00 €
    193.28 € / Pack of 100
    Save 11.72 €
    5% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The risk of cross contamination is eliminated by using Blue SteriWare™ Scrapers. Cleanroom manufactured and available sterile, these funnels are ideal for scraping and cleaning in the food, cosmetic, and pharmaceutical industry. Blue color allows you to see the scraper if left in product.
  • This range of blue disposable Scrapers are available sterile and non-sterile.
  • They are made from high quality polymers.
  • These scrapers are ideal in the manufacture of creams, ointments and pastes.
  • The bright blue color means they can easily seen if they are left in the product by mistake.
  • SteriWare™ Scrapers are available sterile and non-sterile.
  • Moulded and packed in a cleanroom.
  • Individually bagged.
  • Made form FDA and EU conforming materials.
  • BSE/TSE free.
  • Full Batch Traceability.
  • Available as gamma irradiated sterile (SAL 10-6).
Specifications
Sterility Sterile
Dimensions (L x W) 75 x 75 mm
Material Polypropylene
Packaging Individually Bagged