missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Blunt-End Needles
Sterile, 16 gauge, metal luer lock hub, 1.5 in. long
Brand: STEMCELL Technologies 28120
Additional Details : Weight : 6.76220kg
Description
- Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™)
- The blunt end reduces the risk of needle-stick injuries
- Product is manufactured from non-pyrogenic materials
Specifications
Colony Assay | |
2000/Pk. |