Learn More
STEMCELL Technologies Blunt-End Needles, 16 Gauge
Product Code. 16625655
Encompass
Change view
Click to view available options
Needle Gauge:
16 G
Unit Size:
Pack of 100
Product Code. Needle Gauge Unit Size
16625655 16 G Pack of 100
1 options
Product Code. 16625655 Supplier STEMCELL Technologies Supplier No. 28110
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Sterile, 16 gauge, metal luer lock hub, 1.5 in. long
  • Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™)
  • The blunt end reduces the risk of needle-stick injuries
  • Product is manufactured from non-pyrogenic materials
Specifications
Needle Gauge 16 G
For Use With (Application) Colony Assay
Quantity 100/Pk.