Learn More
BMP-15/GDF-9B Rat anti-Human, Clone: 317809, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2925-SP
Encompass
Product Code. 15755764
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, Immunohistochemistry.