missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%

Product Code. 15433287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433287 1 g 1g
11471030 5 g 5g
11481030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433287 Supplier Thermo Scientific Alfa Aesar Supplier No. B25421.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 89524-98-1 | C10H20NO7P | 297.24 g/mol

It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89524-98-1
Molecular Formula C10H20NO7P
Molecular Weight (g/mol) 297.24
MDL Number MFCD02178893
InChI Key LJHAPRKTPAREGO-ZETCQYMHSA-N
Synonym methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
PubChem CID 397380
IUPAC Name methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 297.24
Percent Purity 95%
Chemical Name or Material (+/-)-Boc-alpha-phosphonoglycine trimethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.