missing translation for 'onlineSavingsMsg'
Learn More

(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%

Product Code. 11905501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11905501 5 g 5g
11661329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905501 Supplier Thermo Scientific Acros Supplier No. 439210050
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 89524-98-1 | C10H20NO7P | 297.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89524-98-1
Molecular Formula C10H20NO7P
Molecular Weight (g/mol) 297.24
MDL Number MFCD02178893
InChI Key LJHAPRKTPAREGO-ZETCQYMHSA-N
Synonym methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
PubChem CID 397380
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 69.7°C to 71.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Formula Weight 297.25
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material (±)-BOC-alpha-Phosphonoglycine trimethyl ester
Show More Show Less
compliance-icons
Product Identifier
  • (±)-Boc-alpha-Phosphonoglycine trimethyl ester
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (±)-Boc-alpha-Phosphonoglycine trimethylester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.