missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-Alanine, 99+%

Product Code. 10247020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10247020 5 g Glass Bottle 5g
10666174 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10247020 Supplier Thermo Scientific Acros Supplier No. 275720050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7764-95-6 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7764-95-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00063123
InChI Key QVHJQCGUWFKTSE-RXMQYKEDSA-N
Synonym boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 637606
SMILES C[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 81°C to 84°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CH3)3COC(O)nHC(CH3)COOH
Beilstein 04, 2507
Specific Rotation +25° to +27° (20°C, 589nm) (c=2, CH3COOH)
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material BOC-D-Alanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.