missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Aspartic acid 4-benzylester, 99+%, Thermo Scientific™

Product Code. 10471671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10471671 5 g Glass Bottle 5g
10636924 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10471671 Supplier Thermo Scientific Acros Supplier No. 275520050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7536-58-5
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00065564
InChI Key SOHLZANWVLCPHK-LBPRGKRZSA-N
Synonym boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
PubChem CID 1581888
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 98°C to 102°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula C6H5CH2OCOCH2CH(COOH)nHCOOC(CH3)3
Specific Rotation -18.5° to -21° (20°C, 589nm) (c=2, DMF)
Quantity 5 g
Formula Weight 323.34
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material BOC-L-Aspartic acid 4-benzylester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.