missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Aspartic acid 4-methyl ester, 98%

Product Code. 10515154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10515154 5 g 5g
10366644 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 10515154

Brand: Thermo Scientific Acros 426230050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 59768-74-0 | C10H17NO6 | 247.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59768-74-0
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.25
MDL Number MFCD00078971
InChI Key WFPSMPYVXFVVFA-UHFFFAOYNA-N
Synonym boc-asp ome-oh, boc-l-aspartic acid 4-methyl ester, s-2-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid, boc-asp ome-oh.dcha, s-2-tert-butoxycarbonylamino-4-methoxy-4-oxobutanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-tert-butoxy carbonylamino-3-methoxycarbonyl propanoic acid, boc-asp ome, pubchem12100
PubChem CID 10977771
SMILES COC(=O)CC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation −5° to −7° (20°C, 589nm) (c=2, MeOH)
IUPAC Name (2S)-4-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Quantity 5 g
Formula Weight 247.25
Percent Purity ≥97.5% (Acid)
Physical Form Solution
Chemical Name or Material BOC-L-Aspartic acid 4-methyleester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.