missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Phenylalanine, 99+%

Product Code. 10303822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10303822 5 g Glass Bottle 5g
10368960 25 g Glass Bottle 25g
10599253 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10303822 Supplier Thermo Scientific Acros Supplier No. 275640050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85.0°C to 88.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula C6H5CH2CH(COOH)NHCOOC(CH3)3
Beilstein 14, 1595
Specific Rotation + 25.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and methylene chloride, insoluble in aliphatic hydrocarbons
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Quantity 5 g
Formula Weight 265.3
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-L-Phenylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.