missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-alpha-phenylglycine, 99%

Product Code. 10595144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10595144 1 g Glass Bottle 1g
10386594 5 g Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10595144

Brand: Thermo Scientific Acros 398160010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2900-27-8 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2900-27-8
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065588
InChI Key HOBFSNNENNQQIU-JTQLQIEISA-N
Synonym boc-phg-oh, boc-l-phenylglycine, s-2-tert-butoxycarbonyl amino-2-phenylacetic acid, boc-l-alpha-phenylglycine, boc-l-phg-oh, boc-l-a-phenylglycine, n-boc-l-alpha-phenylglycine, n-boc-l-2-phenylglycine, s-boc-l-phenylglycine, n-boc-l-phenylglycine
PubChem CID 11010409
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 88°C to 91°C
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 251.28
Percent Purity 99%
Chemical Name or Material BOC-L-α-phenylglycine
compliance-icons
Product Identifier
  • BOC-L-alpha-phenylglycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.