missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Proline, 99+%

Product Code. 10378960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10378960 25 g Glass Bottle 25g
10016153 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10378960 Supplier Thermo Scientific Acros Supplier No. 275650250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15761-39-4 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15761-39-4
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00037324
InChI Key ZQEBQGAAWMOMAI-ZETCQYMHSA-N
Synonym boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro
PubChem CID 85083
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 133.0°C to 136.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Beilstein 22, 35
Specific Rotation − 60.50
Water 0.5% max. (K.F.)
Quantity 25 g
Formula Weight 215.25
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material BOC-L-Proline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.