missing translation for 'onlineSavingsMsg'
Learn More

Boc-L-Valine, TRC

Product Code. 30348468
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348468 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348468 Supplier Toronto Research Chemicals Supplier No. TRCB673490100G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-L-Valine
CAS 13734-41-3
Molecular Formula C10H19NO4
Quantity 100 g
Synonym N-[(1,1-dimethylethoxy)carbonyl]-L-valine, (2S)-2-(tert-Butoxycarbonylamino)-3-methylbutanoic Acid, (2S)-2-[[(1,1-Dimethylethoxy)carbonyl]amino]-3-methylbutanoic Acid, (S)-2-(tert-Butoxycarbonylamino)-3-methylbutanoic Acid, (S)-N-tert-Butoxycarbonylvaline, BOC-valine, Boc-L-valine, N-BOC-L-valine, N-BOC-valine, N-tert-Butoxycarbonyl-L-valine, N-tert-Butoxycarbonylvaline, N-tert-Butyloxycarbonyl-L-valine, N-tert-Butyloxycarbonylvaline, N-alpha-tert-Butoxycarbonyl-L-valine, NSC 197197, t-Butoxycarbonyl-L-valine, tert-Butoxycarbonyl-L-valine
InChI Formula InChI=1S/C10H19NO4/c1-6(2)7(8(12)13)11-9(14)15-10(3,4)5/h6-7H,1-5H3,(H,11,14)(H,12,13)/t7-/m0/s1
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Molecular Weight (g/mol) 217.26
Formula Weight 217.1314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.