missing translation for 'onlineSavingsMsg'
Learn More

Boc-ON, 98+%

Product Code. 11403158
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403158 5 g 5g
11413158 25 g 25g
11423158 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403158 Supplier Thermo Scientific Alfa Aesar Supplier No. A18906.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 58632-95-4 | C13H14N2O3 | 246.266 g/mol

Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

Solubility
Soluble in ethyl acetate, ether and benzene. Insoluble in water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58632-95-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.266
MDL Number MFCD00001863
InChI Key QQWYQAQQADNEIC-PTNGSMBKSA-N
Synonym 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
PubChem CID 5868400
IUPAC Name tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
SMILES CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1

Specifications

Melting Point 86°C to 89°C
Quantity 5 g
UN Number UN3439
Beilstein 2217296
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,ether and benzene. Insoluble in water.
Formula Weight 246.27
Percent Purity ≥98%
Chemical Name or Material Boc-ON

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.