missing translation for 'onlineSavingsMsg'
Learn More

BOC-S-trityl-L-cysteine, 99%

Product Code. 10786465 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10786465 1 g Glass Bottle 1g
10603025 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786465 Supplier Thermo Scientific Acros Supplier No. 399060010
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21947-98-8 | C27H29NO4S | 463.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.592
MDL Number MFCD00038251
InChI Key JDTOWOURWBDELG-QHCPKHFHSA-N
Synonym boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
PubChem CID 11167161
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 463.6
Chemical Name or Material BOC-S-trityl-L-cysteine

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.