missing translation for 'onlineSavingsMsg'
Learn More

BOC-Sarcosine, 99+%

Product Code. 10771331 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10771331

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13734-36-6 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-36-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037795
InChI Key YRXIMPFOTQVOHG-UHFFFAOYSA-N
Synonym boc-sar-oh, boc-sarcosine, n-boc-sarcosine, glycine, n-1,1-dimethylethoxy carbonyl-n-methyl, n-tert-butoxycarbonyl-n-methylglycine, t-boc-sarcosine, n-t-boc-sarcosine, 2-tert-butoxy carbonyl methyl amino acetic acid, n-tert-butoxycarbonylsarcosine, tert-butoxycarbonyl methyl amino acetic acid
PubChem CID 83692
IUPAC Name 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(C)CC(=O)O

Specifications

Melting Point 86°C to 90°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass Bottle
Linear Formula (CH3)3COCON(CH3)CH2COOH
Solubility Information Solubility in water: slightly soluble
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material BOC-Sarcosine, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.