missing translation for 'onlineSavingsMsg'
Learn More

BOC-trans-4-hydroxy-L-proline, 98%, ACROS Organics™

Product Code. 10577020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10577020 25 g Glass bottle 25g
10351150 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10577020 Supplier Thermo Scientific Acros Supplier No. 435480250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13726-69-7
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD00053370
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 88804
IUPAC Name 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point 121°C to 125°C
Color White
Elemental Analysis 50.6 to 53.2% C, 5.9 to 6.2% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Specific Rotation -51.5° to -55.5° (20°C, 589nm) (c=1 in DMF)
Formula Weight 231.25
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material BOC-trans-4-Hydroxy-L-proline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.