missing translation for 'onlineSavingsMsg'
Learn More

Boc-trans-Hydroxy-L-proline, TRC

Product Code. 30403515
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403515 1 g -
30393272 5 g 5g
30401607 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403515 Supplier Toronto Research Chemicals Supplier No. TRCH9524951G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-69-7
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
Synonym (2S-trans)-4-Hydroxy-1,2-Pyrrolidinedicarboxylic Acid 1-(1,1-Dimethylethyl) Ester, L-4-Hydroxy-1,2-pyrrolidinedicarboxylic Acid 1-tert-Butyl Ester, (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic Acid, (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic Acid, (2S,4R)-4-Hydroxy-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic Acid, (2S,4R)-4-Hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic Acid, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl Ester, (4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, 1-(tert-Butoxycarbonyl)-(2S,4R)-4-hydroxypyrrolidine-2-carboxylic Acid, Boc-L-trans-gamma-hydroxyproline, Boc-trans-Hydroxy-L-proline, N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, N-(tert-Butoxycarbonyl)hydroxy-L-proline, N-(tert-Butoxycarbonyl)hydroxyproline, N-(tert-Butyloxycarbonyl)-4-hydroxyproline, N-Boc-trans-4-hydroxy-L-proline, N-tert-Butoxycarbonyl-(4R)-4-hydroxy-L-proline, NSC 343720, trans-1-(tert-Butoxycarbonyl)-4-hydroxy-L-proline
IUPAC Name (2S,4R)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C[C@H](O)C[C@H]1C(=O)O

Specifications

InChI Formula InChI=1S/C10H17NO5/c1-10(2,3)16-9(15)11-5-6(12)4-7(11)8(13)14/h6-7,12H,4-5H2,1-3H3,(H,13,14)/t6-,7+/m1/s1
Quantity 1 g
Formula Weight 231.1107
Shipping Condition Room Temperature
Chemical Name or Material Boc-trans-Hydroxy-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.